Learn More
Fluidigm Corp Anti-APC (APC003)-176Yb 100 Tests
SDP
Supplier:  Fluidigm Corp 3176007B
Encompass
The APC003 antibody reacts with both allophycocyanin (APC) alone and APC conjugated with antibody or streptavidin, as well as APC tandem dyes such as APC-Cy5.5 and APC-Cy7.
Catalog No. 50-196-3424