Learn More
Medchemexpress LLC HY-W016289 100mg Medchemexpress, Antiarol CAS:642-71-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW016289100MG
Encompass_Preferred
Medchemexpress, HY-W016289 100mg Antiarol CAS:642-71-7 Antiarol (3,4,5-Trimethoxyphenol) is a natural compound isolated from Salmaliamalabaricum. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4615