Learn More
Medchemexpress LLC Antibacterial agent 18 | 1239602-35-7 | 98.3% | 605.63 g/mol | C39H27NO6 | 1 ML
SDP
Catalog No. 5000210807
Encompass_Preferred
Antibacterial agent 18 is a multi-arm aggregation-induced emission (AIE) antibacterial small molecule derived from patent CN110123801A (compound 23). It is intended for research use and can be supplied as a solid or as a 10 mM solution in DMSO for antibacterial screening and microbiology studies.
  • Multi-arm AIE antibacterial small molecule with activity against Gram-positive and Gram-negative bacteria.
  • Available as a 10 mM solution in DMSO (1 mL) or as solid in multiple mg and g sizes.
  • High purity: 98.3% (as supplied).
  • Supplied with product data sheet and certificate of analysis for quality assurance.
  • Suitable for antibacterial screening and microbiology research applications.
Catalog No. 50-002-10807 Supplier Medchemexpress LLC Supplier No. HYW07464810MM/1ML
Antibacterial agent 18 is a multi-arm aggregation-induced emission (AIE) antibacterial small molecule derived from patent CN110123801A (compound 23). It is intended for research use and can be supplied as a solid or as a 10 mM solution in DMSO for antibacterial screening and microbiology studies.
  • Multi-arm AIE antibacterial small molecule with activity against Gram-positive and Gram-negative bacteria.
  • Available as a 10 mM solution in DMSO (1 mL) or as solid in multiple mg and g sizes.
  • High purity: 98.3% (as supplied).
  • Supplied with product data sheet and certificate of analysis for quality assurance.
  • Suitable for antibacterial screening and microbiology research applications.