Learn More
GenDEPOT ANTIBIOTIC-ANTIMYCOTIC,100ML
Catalog No. 501015476
Encompass_Preferred
Antibiotic-Antimycotic is used to prevent bacterial and fungal contamination. Contains 10,000 units of penicillin (base), 10,000 ug of streptomycin (base), and 25 ug of amphotericin B/ml. Antibiotic-Antimycotic(100X), 100ml
Catalog No. 50-101-5476 Supplier GenDEPOT Supplier No. CA002010
Antibiotic-Antimycotic is used to prevent bacterial and fungal contamination. Contains 10,000 units of penicillin (base), 10,000 ug of streptomycin (base), and 25 ug of amphotericin B/ml. Antibiotic-Antimycotic(100X), 100ml