Learn More
Alkali Scientific Antibiotic Assay Medium 32, 500mL Bottle, 1/Pk
SDP
Catalog No. 5000662664
Encompass_Preferred
Antibiotic Assay Medium 32 is a specialized nutrient medium designed for microbiological potency assays of antibiotics such as dihydrostreptomycin and vancomycin. Each 500 mL bottle contains a carefully balanced blend of beef extract, peptones, and yeast extract prepared to meet USP and FDA specifications. The formulation is adjusted to a pH of approximately 6.6 ± 0.2 to create optimal growth conditions for assay organisms including Klebsiella pneumoniae and Enterococcus hirae. This medium is widely used in pharmaceutical quality control laboratories performing turbidimetric or plate based assays. Its standardized nutrient composition ensures consistent bacterial growth and reproducible results, which are essential for generating accurate calibration curves and determining antibiotic concentration in samples while maintaining compliance with regulatory testing protocols.
Catalog No. 50-006-62664 Supplier Alkali Scientific Supplier No. AAMB32
Antibiotic Assay Medium 32 is a specialized nutrient medium designed for microbiological potency assays of antibiotics such as dihydrostreptomycin and vancomycin. Each 500 mL bottle contains a carefully balanced blend of beef extract, peptones, and yeast extract prepared to meet USP and FDA specifications. The formulation is adjusted to a pH of approximately 6.6 ± 0.2 to create optimal growth conditions for assay organisms including Klebsiella pneumoniae and Enterococcus hirae. This medium is widely used in pharmaceutical quality control laboratories performing turbidimetric or plate based assays. Its standardized nutrient composition ensures consistent bacterial growth and reproducible results, which are essential for generating accurate calibration curves and determining antibiotic concentration in samples while maintaining compliance with regulatory testing protocols.