Learn More
HIMEDIA Antibiotic Assay Medium No. 19, M101, Microbiology Culture Media, Dehydrated Culture Media, Assay Agar, Antifungal Potency Assay, Saccharomyces cerevisiae, Powder, 500 g
Catalog No. NC4013668
Encompass_Preferred
Antibiotic Assay Medium No. 19 is used for the microbiological assay of the antifungal agents Amphotericin B, Candicidin and Nystatin, with Saccharomyces cerevisiae as the test organism. Peptone, yeast extract, HM peptone, dextrose and sodium chloride in a firm agar base, final pH 6.1, give uniform lawns and clearly defined zones of inhibition for accurate potency determination by the agar diffusion method. A reference medium for pharmaceutical antibiotic assay laboratories. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013668 Supplier HIMEDIA Supplier No. M101500G
Antibiotic Assay Medium No. 19 is used for the microbiological assay of the antifungal agents Amphotericin B, Candicidin and Nystatin, with Saccharomyces cerevisiae as the test organism. Peptone, yeast extract, HM peptone, dextrose and sodium chloride in a firm agar base, final pH 6.1, give uniform lawns and clearly defined zones of inhibition for accurate potency determination by the agar diffusion method. A reference medium for pharmaceutical antibiotic assay laboratories. Dehydrated powder, store in a cool dry place. Packaged as 500 g.