Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Antibiotic Disk Dispenser
SCI_ED

Catalog No. S28516 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S28516 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28516 Supplier Innovating Science™ Supplier No. XD1360
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For 50 disk cartridge.

  • Used with Innovating Science Antibiotic disks:
  • Ampicillin 10mcg (XD1361)
  • Chloramphenicol 30mcg (XD1362)
  • Gentamicin 10mcg (XD1363)
  • Penicillin 10units (XD1364)
  • Streptomycin 10mcg (XD1365)
  • Tetracycline 30mcg (XD1366)
  • Blank Sterile disks (XD1368)

Specifications

Grade 7 to 14
Format Kit
Product Type Antibiotic Disk Dispenser
Diameter (English) Ring 0.25 in.
Quantity 250 Disks (5 x 50/Pk.)
Color White
Languages English
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.