Learn More
STEMCELL Technologies Mitomycin C, Size: 50 mg
SDP
Catalog No. NC2188023
Encompass
Inhibit DNA synthesis and mutagenesis, stimulate genetic recombination, and induce DNA repair (Tomasz. Chem Biol, 1995) with Mitomycin C. This antibiotic, which acts as a double-stranded DNA alkylating agent, covalently cross links DNA, inhibiting DNA synthesis and cell proliferation. It acts by way of reductive activation either through low pH or NAD(P)H:quinone oxidoreductase (DT-diaphorase) or NADH cytochrome c reductase (Mao et al. Chem Biol, 1999; Cummings et al. Biochem Pharmacol, 1998). Mitomycin C can mitotically inactivate mouse embryonic fibroblasts (MEFs) for use as feeder cell layers in embryonic stem cell co-culture systems (Bryja et al. Nat Protoc, 2006). Mitomycin C is supplied as a crystalline solid with ≥ 98% purity; Molecular weight 334.3 g/mol; CAS number 50-07-7. Visit STEMCELL.com for more information and related products.
This item is not returnable. View return policy
Catalog No. NC2188023 Supplier STEMCELL Technologies Supplier No. 1001048
Inhibit DNA synthesis and mutagenesis, stimulate genetic recombination, and induce DNA repair (Tomasz. Chem Biol, 1995) with Mitomycin C. This antibiotic, which acts as a double-stranded DNA alkylating agent, covalently cross links DNA, inhibiting DNA synthesis and cell proliferation. It acts by way of reductive activation either through low pH or NAD(P)H:quinone oxidoreductase (DT-diaphorase) or NADH cytochrome c reductase (Mao et al. Chem Biol, 1999; Cummings et al. Biochem Pharmacol, 1998). Mitomycin C can mitotically inactivate mouse embryonic fibroblasts (MEFs) for use as feeder cell layers in embryonic stem cell co-culture systems (Bryja et al. Nat Protoc, 2006). Mitomycin C is supplied as a crystalline solid with ≥ 98% purity; Molecular weight 334.3 g/mol; CAS number 50-07-7. Visit STEMCELL.com for more information and related products.