Learn More
AdipoGen Antibiotic L-696 474
SDP
Catalog No. 502561986
Encompass_Preferred
Chemical. CAS 141994-72-1. Formula C30H39NO4. MW 477.6. Isolated from Hypoxylon fragiforme. Natural cytochalasin derivative. Antiviral. Inhibitor of HIV-1 protease. Antifungal activity.
Catalog No. 50-256-1986 Supplier AdipoGen Supplier No. BVT0331C500
Chemical. CAS 141994-72-1. Formula C30H39NO4. MW 477.6. Isolated from Hypoxylon fragiforme. Natural cytochalasin derivative. Antiviral. Inhibitor of HIV-1 protease. Antifungal activity.