Learn More
TOKU-E Polymyxin B ReadyMade™ Solution, 1405-20-5, 10 ML
Catalog No. 502551759
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 20 mg/ml in water. Useful in microbiology, plant biology.
Catalog No. 50-255-1759 Supplier TOKU-E Supplier No. P17410ML
Antibiotic, ReadyMade™, sterile-filtered, 20 mg/ml in water. Useful in microbiology, plant biology.