Learn More
TOKU-E Kanamycin ReadyMade™ Solution, 25389-94-0, 100 ML
Catalog No. 502551781
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 10 mg/ml in 0.9% NaCl. Useful in cancer research, cell culture, microbiology.
Catalog No. 50-255-1781 Supplier TOKU-E Supplier No. K036100ML
Antibiotic, ReadyMade™, sterile-filtered, 10 mg/ml in 0.9% NaCl. Useful in cancer research, cell culture, microbiology.