Learn More
STEMCELL Technologies Bleomycin (Sulfate), Size: 5 mg
SDP
Catalog No. 502069358
Encompass
Bleomycin is a glycopeptide-derived antibiotic, isolated from the bacterium Streptomyces verticillus, that creates DNA strand scission and exhibits antitumor activity against squamous cell carcinomas and malignant lymphomas (Galm et al.). Bleomycin induces lung injury leading to acute inflammatory response and pulmonary fibrosis (Kulkarni et al.). This product is supplied as the sulfate salt of the molecule.CANCER RESEARCH· Induces single- and double-strand DNA breaks in tumor cells (Hecht).DISEASE MODELING· Induces lung fibrosis in mice (Kulkarni et al.).
Catalog No. 50-206-9358 Supplier STEMCELL Technologies Supplier No. 1000550
Bleomycin is a glycopeptide-derived antibiotic, isolated from the bacterium Streptomyces verticillus, that creates DNA strand scission and exhibits antitumor activity against squamous cell carcinomas and malignant lymphomas (Galm et al.). Bleomycin induces lung injury leading to acute inflammatory response and pulmonary fibrosis (Kulkarni et al.). This product is supplied as the sulfate salt of the molecule.CANCER RESEARCH· Induces single- and double-strand DNA breaks in tumor cells (Hecht).DISEASE MODELING· Induces lung fibrosis in mice (Kulkarni et al.).