Learn More
Antibiotics - IR Identification PT, MilliporeSigma™ Supelco™
Catalog No. 111004318 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4 x 1 g
Catalog No. Quantity
11-100-4318 4 x 1 g
1 options
Catalog No. 11-100-4318 Supplier MilliporeSigma Supelco Supplier No. PEP0034X1G
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY
Specifications
Description IR Identification PT
Packaging Ampule
Type Proficiency Testing Material
Quantity 4 x 1 g