Learn More
Fluidigm Corp Anti-Biotin (1D4-C5)-165Ho 100 Tests
SDP
Supplier:  Fluidigm Corp 3165012B
Encompass
The 1D4-C5 monoclonal antibody can be used as a second-step reagent to detect cells stained with biotinylated primary antibodies.
Catalog No. 50-196-3152