Learn More
ACROBiosystems Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1
Supplier:  ACROBiosystems FM3Y451000TESTS
Encompass_Preferred
Antibody;1000TESTS;IgG1/kappa;Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.;Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with FMC63 scFv.; ;2e5 of FMC63 scFv-based anti-CD19 CAR-293 cells were stained with 100 µL of the working solution of Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Cat. No. FM3-Y45) and isotype control respectively, washed and then followed by PE anti-mouse IgG1 Antibody and analyzed with flow cytometry (QC tested).
Catalog No. 50-201-9761
Add to Cart
Request a Quote