Learn More
ACROBiosystems Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1
Catalog No. 502019534
Encompass_Preferred
Antibody;100UG;Human IgG1/kappa;The cross-reactivity with other coronaviruses has not been tested yet.;Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (NUN-S41) is isolated from a SARS-CoV-2 infected patient and is recombinantly produced from human 293 cells (HEK293).;This protein . ;Immobilized SARS-CoV-2 Nucleocapsid protein, His Tag (Cat. No. NUN-C5227) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41) with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-201-9534 Supplier ACROBiosystems Supplier No. NUNS41100UG
Antibody;100UG;Human IgG1/kappa;The cross-reactivity with other coronaviruses has not been tested yet.;Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (NUN-S41) is isolated from a SARS-CoV-2 infected patient and is recombinantly produced from human 293 cells (HEK293).;This protein . ;Immobilized SARS-CoV-2 Nucleocapsid protein, His Tag (Cat. No. NUN-C5227) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41) with a linear range of 0.2-2 ng/mL (QC tested).