Antibody;100UG;Mouse IgG1/kappa;The product is specific for Human BCMA Protein extracellular, and can inhibit the interaction between BCAM and BAFF (IC50= 4.877 μg/mL), BCAM and APRIL (IC50= 4.624 μg/mL). It also has cross reactivity with Cynomolgus & Mouse BCMA protein.;Anti-BCMA Neutralizing Antibody, Mouse IgG1 (BCA-M43) recognizes the Human BCMA Protein extracellular and can inhibit the interaction between BCMA and BAFF/APRIL. It is expressed from human 293 cells (HEK293).; ;Immobilized Human BCMA, His Tag (Cat. No. BCA-H522y) at 2 μg/mL (100 μL/well) can bind Anti-BCMA Neutralizing Antibody, Mouse IgG1 (Clone: AM43, Cat. No. BCA-M43) with a linear range of 0.256-6.4 ng/mL (QC tested).;Immobilized Human BCMA, His Tag (Cat. No. BCA-H522y) at 2 μg/mL (100 μL/well) can bind pre-mixed increasing concentrations of Anti-BCMA Neutralizing Antibody, Mouse IgG1 (Clone: AM43, Cat. No. BCA-M43) and 0.01 μg/mL (100 μL/well) Biotinylated Human BAFF, His,Avitag (Cat. No. BAF-H82Q2) with a half m