Learn More
Cayman Chemical 11-oxo MGV, Purity: ≥95%, CAS: 126105-11-1, 1 mg
SDP
Catalog No. 502987112
Encompass_Preferred
A sweet tasting cucurbitane glycoside antioxidant (EC50s = 4.79 and 16.52 µg/ml against O2- and OH-, respectively); inhibits carcinogenesis in murine models
Catalog No. 50-298-7112 Supplier Cayman Chemical Supplier No. 224161 MG
A sweet tasting cucurbitane glycoside antioxidant (EC50s = 4.79 and 16.52 µg/ml against O2- and OH-, respectively); inhibits carcinogenesis in murine models