Learn More
Cayman Chemical ANTIBODY 13 S-HODE 100ug
SDP
Catalog No. 502916348
Encompass_Preferred
13(S)-HODE is produced by incubation of linoleic acid with plant and mammalian LOs. It has been shown to inhibit the adhesion of tumor cells to the endothelium at concentrations around 1 µM,{592,880} and down regulates the expression of the IRGpIIb/IIIa receptor.{591}
Catalog No. 50-291-6348 Supplier Cayman Chemical Supplier No. 38610100 UG
13(S)-HODE is produced by incubation of linoleic acid with plant and mammalian LOs. It has been shown to inhibit the adhesion of tumor cells to the endothelium at concentrations around 1 µM,{592,880} and down regulates the expression of the IRGpIIb/IIIa receptor.{591}