Learn More
Cayman Chemical ANTIBODY 15 S-HpEDE 250ug
SDP
Catalog No. 502945281
Encompass_Preferred
15(S)-HpEDE is a monohydroperoxy PUFA produced by the action of 15-LO on eicosadienoic acid. Although the biological activities of 15(S)-HpEDE have not been well characterized, it is expected to behave similarly to 15(S)-HpETE (Catalog No. 44720).
Catalog No. 50-294-5281 Supplier Cayman Chemical Supplier No. 47720250 UG
15(S)-HpEDE is a monohydroperoxy PUFA produced by the action of 15-LO on eicosadienoic acid. Although the biological activities of 15(S)-HpEDE have not been well characterized, it is expected to behave similarly to 15(S)-HpETE (Catalog No. 44720).