Learn More
Cayman Chemical 1O14, Purity: ≥95%, CAS: 3117189-57-5, 25 mg
SDP
Catalog No. 502916322
Encompass_Preferred
An ionizable cationic lipid; has been used in combination with other lipids in the formation of LNPs; 1O14-containing LNPs have been used for the delivery of IL-1β siRNA to induce gene silencing and hepatoprotective effects in a mouse model of acute liver injury
Catalog No. 50-291-6322 Supplier Cayman Chemical Supplier No. 3815025 MG
An ionizable cationic lipid; has been used in combination with other lipids in the formation of LNPs; 1O14-containing LNPs have been used for the delivery of IL-1β siRNA to induce gene silencing and hepatoprotective effects in a mouse model of acute liver injury