Learn More
Cayman Chemical 2-NP-AHD, Purity: ≥95%, CAS: 623145-57-3, 25 mg
SDP
Catalog No. 502912044
Encompass_Preferred
A nitrophenyl derivative of AHD; has been used in immunoassays for the detection of AHD in honey
Catalog No. 50-291-2044 Supplier Cayman Chemical Supplier No. 3650025 MG
A nitrophenyl derivative of AHD; has been used in immunoassays for the detection of AHD in honey