Learn More
Cayman Chemical 2-NP-AOZ, Purity: ≥95%, CAS: 19687-73-1, 50 mg
SDP
Catalog No. 502909953
Encompass_Preferred
A derivatized metabolite of furazolidone; formed from tissue-bound AOZ after acid hydrolysis and derivatization with o-NBA; has been used as a marker of furazolidone use in pigs
Catalog No. 50-290-9953 Supplier Cayman Chemical Supplier No. 3557550 MG
A derivatized metabolite of furazolidone; formed from tissue-bound AOZ after acid hydrolysis and derivatization with o-NBA; has been used as a marker of furazolidone use in pigs