Learn More
Cayman Chemical ANTIBODY 20-HETE 50UG
SDP
Supplier:  Cayman Chemical 9003050 UG
Encompass_Preferred
A CYP450 metabolite of arachidonic acid in the renal and cerebral vasculature; exhibits vasoconstrictor activity
Catalog No. 50-289-2737