Learn More
ACROBiosystems FITC-Labeled Anti-Rituximab Antibodies, Mouse IgG1
Supplier:  ACROBiosystems RIBFY35C200UG
Encompass_Preferred
Antibody;200UG;IgG1/kappa;Recognizes Rituximab specifically, no cross reactivity with other humanized antibodies.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Rituximab F(ab')2.;This protein . ;Immobilized Rituximab at 2 μg/mL (100 μL/well) can bind FITC-Labeled Anti-Rituximab Antibodies, Mouse IgG1 (Cat. No. RIB-FY35c) with a linear range of 0.04-0.6 μg/mL (QC tested).;2e5 of Anti-CD20(Rituximab) CAR-293 cells were stained with 100 μL of 3 μg/mL of FITC-labeled Anti-Rituximab Antibodies (Cat. No. RIB-FY35c) and isotype control protein respectively. FITC signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-201-9452