Learn More
ACROBiosystems Biotinylated Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control
Catalog No. 502019430
Encompass_Preferred
Antibody;200UG;Mouse IgG1/kappa;Specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.;Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies. Biotinylated Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control was purified from HEK293 cell culture. It is the Biotin labeled form ofMonoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control (Cat. No. DNP-M1).;This protein . ;2e5 of Anti-CD19 CAR-293 cells were stained with a series of concentrations of Biotinylated Anti-DNP Antibody, Mouse IgG1 (Cat. No. DNP-BM1), washed and then followed by PE-SA and analyzed with FACS (QC tested). ;5e5 of PBMCs were stained with a series of concentrations of Biotinylated Anti-DNP Antibody, Mouse IgG1 (Cat. No. DNP-BM1
Catalog No. 50-201-9430 Supplier ACROBiosystems Supplier No. DNPBM1200UG
Antibody;200UG;Mouse IgG1/kappa;Specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.;Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies. Biotinylated Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control was purified from HEK293 cell culture. It is the Biotin labeled form ofMonoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control (Cat. No. DNP-M1).;This protein . ;2e5 of Anti-CD19 CAR-293 cells were stained with a series of concentrations of Biotinylated Anti-DNP Antibody, Mouse IgG1 (Cat. No. DNP-BM1), washed and then followed by PE-SA and analyzed with FACS (QC tested). ;5e5 of PBMCs were stained with a series of concentrations of Biotinylated Anti-DNP Antibody, Mouse IgG1 (Cat. No. DNP-BM1