Learn More
Cayman Chemical ANTIBODY 22-HDHA 100ug
SDP
Catalog No. 502982288
Encompass_Preferred
An oxidation product of DHA; formed by human CYP4F3B in BTI-TN-5B1-4 microsomes; serum levels increase in humans following dietary DHA supplementation
Catalog No. 50-298-2288 Supplier Cayman Chemical Supplier No. 19321100 UG
An oxidation product of DHA; formed by human CYP4F3B in BTI-TN-5B1-4 microsomes; serum levels increase in humans following dietary DHA supplementation