Learn More
ACROBiosystems FITC-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1
Catalog No. 502019666
Encompass_Preferred
Antibody;25TESTS;IgG1/kappa;Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.;Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with FMC63 scFv. It is the FITC labeled form of Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (FM3-Y45).;This protein . ;2e5 of Anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) FITC-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Cat. No. FM3-FY45) and isotype control respectively. FITC signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-201-9666 Supplier ACROBiosystems Supplier No. FM3FY4525TESTS
Add to Cart
Add to Cart
Antibody;25TESTS;IgG1/kappa;Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.;Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with FMC63 scFv. It is the FITC labeled form of Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (FM3-Y45).;This protein . ;2e5 of Anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) FITC-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Cat. No. FM3-FY45) and isotype control respectively. FITC signal was used to evaluate the binding activity (QC tested).