Learn More
Cayman Chemical 5,15-DPP, Purity: ≥95%, CAS: 22112-89-6, 10 mg
SDP
Catalog No. 502971220
Encompass_Preferred
A cell-permeable porphyrin derivative that selectively binds STAT3 (Kd = 880 nM); inhibits STAT3 dimerization via Src homology 2 (Sh2) domains (IC50 = 280 nM), preventing nuclear translocation and DNA binding
Catalog No. 50-297-1220 Supplier Cayman Chemical Supplier No. 1609010 MG
A cell-permeable porphyrin derivative that selectively binds STAT3 (Kd = 880 nM); inhibits STAT3 dimerization via Src homology 2 (Sh2) domains (IC50 = 280 nM), preventing nuclear translocation and DNA binding