Learn More
Cayman Chemical ANTIBODY 5-Ph-IAA 25mg
SDP
Catalog No. 502916486
Encompass_Preferred
A phenylic substituted indole and ligand used for bump and hole; activates a modified TIR1, which forms a PROTAC-associated complex that targets and degrades RAD21 and induces cell cycle arrest at the G2 or M phase in Arabidopsis at 1 µM; reduces tumor growth via the same process by targeting and degrading BRD4 in an HCT116 colorectal cancer xenograft mouse model at 1, 3, or 10 mg/kg per day, i.p.
Catalog No. 50-291-6486 Supplier Cayman Chemical Supplier No. 3816125 MG
A phenylic substituted indole and ligand used for bump and hole; activates a modified TIR1, which forms a PROTAC-associated complex that targets and degrades RAD21 and induces cell cycle arrest at the G2 or M phase in Arabidopsis at 1 µM; reduces tumor growth via the same process by targeting and degrading BRD4 in an HCT116 colorectal cancer xenograft mouse model at 1, 3, or 10 mg/kg per day, i.p.