Learn More
Cayman Chemical 6-PPD-Q, Purity: ≥95%, CAS: 2754428-18-5, 1 mg
SDP
Catalog No. 502917521
Encompass_Preferred
An oxidized transformation product of 6-PPD; has been detected as a contaminant in atmospheric particles, various aquatic sources, including surface water, stormwater, and wastewater, and road dust, among other media; highly toxic to aquatic organisms, including salmon and zebrafish; induces oxidative stress, DNA damage, inflammation, and reproductive dysfunction in a variety of model organisms
Catalog No. 50-291-7521 Supplier Cayman Chemical Supplier No. 382471 MG
An oxidized transformation product of 6-PPD; has been detected as a contaminant in atmospheric particles, various aquatic sources, including surface water, stormwater, and wastewater, and road dust, among other media; highly toxic to aquatic organisms, including salmon and zebrafish; induces oxidative stress, DNA damage, inflammation, and reproductive dysfunction in a variety of model organisms