Learn More
Cayman Chemical ANTIBODY 6-PPD-Q 25mg
SDP
Catalog No. 502916651
Encompass_Preferred
An oxidized derivative of 6-PPD; toxic to rainbow trout and brook trout (LC50s = 0.59 and 1.96 UG/L, respectively) but not to arctic char and white sturgeon (LC50s = >12.7 UG/L for both); induces cell death and germline DNA damage, as well as decreases the number of mitotic cells, in C. elegans gonads at 10 UG/L; urine levels are increased in pregnant women compared to non-pregnant adults and children
Catalog No. 50-291-6651 Supplier Cayman Chemical Supplier No. 3824725 MG
An oxidized derivative of 6-PPD; toxic to rainbow trout and brook trout (LC50s = 0.59 and 1.96 UG/L, respectively) but not to arctic char and white sturgeon (LC50s = >12.7 UG/L for both); induces cell death and germline DNA damage, as well as decreases the number of mitotic cells, in C. elegans gonads at 10 UG/L; urine levels are increased in pregnant women compared to non-pregnant adults and children