Learn More
Cayman Chemical ANTIBODY 7-PPD-Q 500ug
SDP
Catalog No. 502944326
Encompass_Preferred
A PPD-Q and derivative of 7-PPD; toxic to the bacterium V. fischeri (EC50 = 14.9 mg/L); has been found in PM2.5 samples collected at the roadside in China
Catalog No. 50-294-4326 Supplier Cayman Chemical Supplier No. 40610500 UG
A PPD-Q and derivative of 7-PPD; toxic to the bacterium V. fischeri (EC50 = 14.9 mg/L); has been found in PM2.5 samples collected at the roadside in China