Learn More
Cayman Chemical ANTIBODY 8 S-HEPE 100ug
SDP
Catalog No. 502949499
Encompass_Preferred
8(S)-HEPE is a monohydroxy fatty acid produced by lipoxygenase oxidation of EPA. It acts to promote hatching of barnacle eggs at 10 nM, although it is not clearly identified as the natural egg hatching factor.{4399}
Catalog No. 50-294-9499 Supplier Cayman Chemical Supplier No. 32350100 UG
8(S)-HEPE is a monohydroxy fatty acid produced by lipoxygenase oxidation of EPA. It acts to promote hatching of barnacle eggs at 10 nM, although it is not clearly identified as the natural egg hatching factor.{4399}