Learn More
Cayman Chemical A-196, Purity: ≥98%, CAS: 1982372-88-2, 10 mg
SDP
Catalog No. 502968303
Encompass_Preferred
A selective inhibitor of SUV420H1 and SUV420H2 (IC50s = 25 and 144 nM, respectively) that inhibits the di- and trimethylation of H4K20me in multiple cell lines
Catalog No. 50-296-8303 Supplier Cayman Chemical Supplier No. 1831710 MG
A selective inhibitor of SUV420H1 and SUV420H2 (IC50s = 25 and 144 nM, respectively) that inhibits the di- and trimethylation of H4K20me in multiple cell lines