Learn More
Cayman Chemical A-485, Purity: ≥98% (mixture of isomers), CAS: 1889279-16-6, 10 mg
SDP
Catalog No. 502933131
Encompass_Preferred
A p300/CBP inhibitor (IC50 = 60 nM); decreases H3K27Ac levels in PC3 cells; reduces proliferation of various tumor cell lines; inhibits expression of PSA and H3K27Ac occupancy at the PSA enhancer in LNCaP-FGC cells; reduces tumor volume in a LuCaP-77 mouse xenograft model of castration-resistant prostate cancer at 100 mg twice per day for 21 days
Catalog No. 50-293-3131 Supplier Cayman Chemical Supplier No. 2411910 MG
A p300/CBP inhibitor (IC50 = 60 nM); decreases H3K27Ac levels in PC3 cells; reduces proliferation of various tumor cell lines; inhibits expression of PSA and H3K27Ac occupancy at the PSA enhancer in LNCaP-FGC cells; reduces tumor volume in a LuCaP-77 mouse xenograft model of castration-resistant prostate cancer at 100 mg twice per day for 21 days