Learn More
Cayman Chemical A-740003, Purity: ≥98%, CAS: 861393-28-4, 25 mg
SDP
Catalog No. 502981474
Encompass_Preferred
A selective P2X7 antagonist (IC50s = 40 and 18 nM for human and rat receptors, respectively); blocks agonist-induced IL-1β release and pore formation in differentiated THP-1 cells (IC50s = 156 and 92 nM, respectively); effective in vivo when administered intraperitoneally
Catalog No. 50-298-1474 Supplier Cayman Chemical Supplier No. 2125625 MG
A selective P2X7 antagonist (IC50s = 40 and 18 nM for human and rat receptors, respectively); blocks agonist-induced IL-1β release and pore formation in differentiated THP-1 cells (IC50s = 156 and 92 nM, respectively); effective in vivo when administered intraperitoneally