Learn More
Cayman Chemical A-769662, Purity: ≥98%, CAS: 844499-71-4, 5 mg
SDP
Catalog No. 502952883
Encompass_Preferred
A direct activator of AMPK (EC50 = 116 nM in a cell-free assay); allosterically activates AMPK and inhibits its dephosphorylation on Thr172; activates β1 subunit-containing AMPK heterotrimers; has been used to stimulate CYP450-mediated fatty acid oxidation, inhibit adipocyte differentiation, explore glucose uptake in skeletal muscle, and promote endothelial cell survival during metabolic stress
Catalog No. 50-295-2883 Supplier Cayman Chemical Supplier No. 119005 MG
A direct activator of AMPK (EC50 = 116 nM in a cell-free assay); allosterically activates AMPK and inhibits its dephosphorylation on Thr172; activates β1 subunit-containing AMPK heterotrimers; has been used to stimulate CYP450-mediated fatty acid oxidation, inhibit adipocyte differentiation, explore glucose uptake in skeletal muscle, and promote endothelial cell survival during metabolic stress