Learn More
Cayman Chemical A-771726, Purity: ≥98%, CAS: 163451-81-8, 25 mg
SDP
Catalog No. 502972692
Encompass_Preferred
An active metabolite of leflunomide; inhibits DHODH (IC50 = 0.23 µM); inhibits the production of PGE2 in TNF-α- or IL-1α-stimulated isolated human synoviocytes (IC50s = 7 and 3 μM, respectively); inhibits the proliferation of isolated human PBMCs at 25 and 100 µM; delays disease onset and decreases neurological deficits in a rat model of EAE induced by CFA and M. tuberculosis at 3 and 10 mg/kg
Catalog No. 50-297-2692 Supplier Cayman Chemical Supplier No. 1440425 MG
An active metabolite of leflunomide; inhibits DHODH (IC50 = 0.23 µM); inhibits the production of PGE2 in TNF-α- or IL-1α-stimulated isolated human synoviocytes (IC50s = 7 and 3 μM, respectively); inhibits the proliferation of isolated human PBMCs at 25 and 100 µM; delays disease onset and decreases neurological deficits in a rat model of EAE induced by CFA and M. tuberculosis at 3 and 10 mg/kg