Learn More
Cayman Chemical A-867744, Purity: ≥98%, CAS: 1000279-69-5, 10 mg
SDP
Catalog No. 502948730
Encompass_Preferred
A positive allosteric modulator of α7 nAChRs; selective for α7 nAChRs over α3β2- and α4β2 subunit-containing nAChRs (IC50s = 0.023, 19.5 and 6.4 µM, respectively) and 5-HT3A receptors (Ki = >30 µM); potentiates ACh-induced currents in Xenopus oocytes expressing human α7 nAChRs (EC50 = ~1 µM); improves sensory gating deficits in a paired auditory stimulus paradigm in mice from 0.1-10 μmol/kg, i.p.
Catalog No. 50-294-8730 Supplier Cayman Chemical Supplier No. 3297410 MG
A positive allosteric modulator of α7 nAChRs; selective for α7 nAChRs over α3β2- and α4β2 subunit-containing nAChRs (IC50s = 0.023, 19.5 and 6.4 µM, respectively) and 5-HT3A receptors (Ki = >30 µM); potentiates ACh-induced currents in Xenopus oocytes expressing human α7 nAChRs (EC50 = ~1 µM); improves sensory gating deficits in a paired auditory stimulus paradigm in mice from 0.1-10 μmol/kg, i.p.