Learn More
Cayman Chemical A286982, Purity: ≥98%, CAS: 280749-17-9, 10 mg
SDP
Catalog No. 502938842
Encompass_Preferred
An inhibitor that blocks the integrin-ligand interaction between LFA-1 and ICAM-1 (IC50 = 35 nM); inhibits adhesion of JY-8 cells to ICAM-1-coated microtiter plates (IC50 = 44 nM)
Catalog No. 50-293-8842 Supplier Cayman Chemical Supplier No. 2764610 MG
An inhibitor that blocks the integrin-ligand interaction between LFA-1 and ICAM-1 (IC50 = 35 nM); inhibits adhesion of JY-8 cells to ICAM-1-coated microtiter plates (IC50 = 44 nM)