Learn More
Cayman Chemical AA3-DLin, Purity: ≥95%, CAS: 3027565-91-6, 25 mg
SDP
Catalog No. 502916822
Encompass_Preferred
An ionizable cationic amino lipid (pKa = 5.8); has been used in combination with other lipids in the formation of LNPs for mRNA delivery; LNPs containing AA3-DLin and encapsulating SARS-CoV-2 spike glycoprotein mRNA induce immunogenicity in mice
Catalog No. 50-291-6822 Supplier Cayman Chemical Supplier No. 3790325 MG
An ionizable cationic amino lipid (pKa = 5.8); has been used in combination with other lipids in the formation of LNPs for mRNA delivery; LNPs containing AA3-DLin and encapsulating SARS-CoV-2 spike glycoprotein mRNA induce immunogenicity in mice