Learn More
Cayman Chemical AA92593, Purity: ≥98%, CAS: 457961-34-1, 10 mg
SDP
Catalog No. 502913028
Encompass_Preferred
An OPN4 receptor antagonist (Ki = 16 nM); inhibits light-induced calcium transients in CHO cells expressing human OPN4 (IC50 = 665 nM); reverses light-induced calcium influx in rat ipRGCs at 10 µM; reduces light-induced pupil constriction in mice at 30 mg/kg
Catalog No. 50-291-3028 Supplier Cayman Chemical Supplier No. 3583610 MG
An OPN4 receptor antagonist (Ki = 16 nM); inhibits light-induced calcium transients in CHO cells expressing human OPN4 (IC50 = 665 nM); reverses light-induced calcium influx in rat ipRGCs at 10 µM; reduces light-induced pupil constriction in mice at 30 mg/kg