Learn More
Cayman Chemical AB-928, Purity: ≥98%, CAS: 2239273-34-6, 50 mg
SDP
Catalog No. 502991097
Encompass_Preferred
An adenosine A2A and A2B receptor dual antagonist; inhibits the ability of adenosine to suppress activation of human CD4 or CD8 T cells, as well as monocyte-derived dendritic cells, in vitro; reduces tumor growth in an AT3-OVA syngeneic mouse model alone or in combination with doxorubicin; acts synergistically with an anti-PD-1 antibody to reduce tumor growth in a B16/F10 syngeneic mouse model
Catalog No. 50-299-1097 Supplier Cayman Chemical Supplier No. 3144450 MG
An adenosine A2A and A2B receptor dual antagonist; inhibits the ability of adenosine to suppress activation of human CD4 or CD8 T cells, as well as monocyte-derived dendritic cells, in vitro; reduces tumor growth in an AT3-OVA syngeneic mouse model alone or in combination with doxorubicin; acts synergistically with an anti-PD-1 antibody to reduce tumor growth in a B16/F10 syngeneic mouse model