Learn More
Cayman Chemical AB-ICA, Purity: ≥98%, 5 mg
SDP
Catalog No. 502917017
Encompass_Preferred
An analytical references standard categorized as a precursor in the synthesis of 5-fluoro ABICA, 4-cyano AB-BUTICA, and AB-FUBICA; intended for research and forensic applications
Catalog No. 50-291-7017 Supplier Cayman Chemical Supplier No. 394015 MG
An analytical references standard categorized as a precursor in the synthesis of 5-fluoro ABICA, 4-cyano AB-BUTICA, and AB-FUBICA; intended for research and forensic applications