Learn More
Cayman Chemical Abametapir, Purity: ≥98%, CAS: 1762-34-1, 5 g
SDP
Catalog No. 502988717
Encompass_Preferred
A building block and an insecticide; has been used in the synthesis of lanthanide coordination complexes with antifungal activity against C. albicans and divalent vancomycin derivatives with antibacterial activity against vancomycin-resistant enterococci; ovicidal to DDT- and permethrin-resistant head louse eggs (LC50s = 0.108, 0.121, and 0.16% in isopropanol for 0-2-, 3-5-, and 6-8-day-old eggs, respectively)
Catalog No. 50-298-8717 Supplier Cayman Chemical Supplier No. 306845 G
A building block and an insecticide; has been used in the synthesis of lanthanide coordination complexes with antifungal activity against C. albicans and divalent vancomycin derivatives with antibacterial activity against vancomycin-resistant enterococci; ovicidal to DDT- and permethrin-resistant head louse eggs (LC50s = 0.108, 0.121, and 0.16% in isopropanol for 0-2-, 3-5-, and 6-8-day-old eggs, respectively)