Learn More
Cayman Chemical Ac-ANW-AMC, Purity: ≥95%, 1 mg
SDP
Catalog No. 502934808
Encompass_Preferred
A fluorogenic substrate for the β5i/LMP7 subunit of the 20S proteasome; AMC is released upon enzymatic cleavage by the β5i/LMP7 subunit of the immunoproteasome and its fluorescence can be used to quantify enzyme activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-293-4808 Supplier Cayman Chemical Supplier No. 266401 MG
A fluorogenic substrate for the β5i/LMP7 subunit of the 20S proteasome; AMC is released upon enzymatic cleavage by the β5i/LMP7 subunit of the immunoproteasome and its fluorescence can be used to quantify enzyme activity; ex/em = 340-360/440-460 nm, respectively