Learn More
Cayman Chemical Acifran, Purity: ≥98%, CAS: 72420-38-3, 10 mg
SDP
Catalog No. 502947844
Encompass_Preferred
A niacin receptor agonist; binds to HCA2 (IC50 = 1.12 µM); induces ERK1/2 phosphorylation in CHO-K1 cells expressing HCA2 or HCA3 from 0.01-10 µM; reduces serum triglyceride and LDL levels and increases the ratio of HDL to total cholesterol in hyperlipidemic rats at 1 mg/kg; reduces serum triglyceride levels in a rat model of STZ-induced diabetes
Catalog No. 50-294-7844 Supplier Cayman Chemical Supplier No. 3455610 MG
A niacin receptor agonist; binds to HCA2 (IC50 = 1.12 µM); induces ERK1/2 phosphorylation in CHO-K1 cells expressing HCA2 or HCA3 from 0.01-10 µM; reduces serum triglyceride and LDL levels and increases the ratio of HDL to total cholesterol in hyperlipidemic rats at 1 mg/kg; reduces serum triglyceride levels in a rat model of STZ-induced diabetes