Learn More
Cayman Chemical Acumapimod, Purity: ≥98%, CAS: 836683-15-9, 25 mg
SDP
Catalog No. 502934961
Encompass_Preferred
An inhibitor of p38α MAPK (IC50 = 50 = 0.3 mg/kg)
Catalog No. 50-293-4961 Supplier Cayman Chemical Supplier No. 2583225 MG
An inhibitor of p38α MAPK (IC50 = 50 = 0.3 mg/kg)