Learn More
Cayman Chemical ANTIBODY ADB-ICA 5mg
SDP
Catalog No. 502943562
Encompass_Preferred
An analytical reference standard categorized as a precursor in the synthesis of synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-294-3562 Supplier Cayman Chemical Supplier No. 399525 MG
An analytical reference standard categorized as a precursor in the synthesis of synthetic cannabinoids; intended for research and forensic applications